Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CYN 154806

Click to view available options
Quantity:
1 mg
Description
CYN 154806 is a potent and selective somatostatin sst2 receptor antagonist. pIC50 values are 8.58, 5.41, 6.07, 5.76 and 6.48 for human recombinant sst2, sst1, sst3, sst4 and sst5 receptors respectively.
Specifications
Specifications
Purity | 0.95 |
Content And Storage | Store at -20°C |
Description | CYN 154806 |
Molecular Weight (g/mol) | 1197.35 |
Product Type | CYN 154806 |
Quantity | 1 mg |
Sequence | Ac-(4-Nitro)Phe-D-Cys-Tyr-D-Trp-Lys-Thr-Cys-D-Tyr-NH2 (Disulfide bridge: 2 - 7) |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction