Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cynomolgus Monkey B7-H6 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10127R100UG
This item is not returnable.
View return policy
Description
B7-H6 Monoclonal specifically detects B7-H6 in Cynomolgus Monkey samples. It is validated for Flow Cytometry.Specifications
B7-H6 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
Mouse | |
100 μg | |
374383 | |
Cynomolgus Monkey | |
IgG2b |
Flow Cytometry | |
1002932 | |
Flow Cytometry 0.25-1 μg/106 cells | |
B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1 | |
Human embryonic kidney cell, HEK293-derived cynomolgus Monkey B7-H6, Asp25-Ser262 | |
Primary | |
Detects Cynomolgus Monkey B7-H6 in direct ELISAs. In direct ELISAs, no cross-reactivity with human B7-H6 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction