Learn More
Cynomolgus Monkey B7-H6 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB10127S100UG
Catalog No. FB10127S100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
B7-H6 Monoclonal specifically detects B7-H6 in Cynomolgus Monkey samples. It is validated for Flow Cytometry.
Specifications
Specifications
B7-H6
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects Cynomolgus Monkey B7-H6 in direct ELISAs. In direct ELISAs, no cross-reactivity with human B7-H6 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b
Flow Cytometry
1002932
Flow Cytometry 0.25-1 ug/10^6 cells
B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Human embryonic kidney cell, HEK293-derived cynomolgus Monkey B7-H6, Asp25-Ser262
100 μg
374383.0
Cynomolgus Monkey
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.