Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYorf15B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548931
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548931 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548931 Supplier Invitrogen™ Supplier No. PA548931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYorf15B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CYorf15B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TXLNGY
Gene Accession No. Q9BZA5
Gene Alias CYorf15A; CYorf15B; taxilin gamma pseudogene, Y-linked; TXLNG2P; TXLNGY
Gene Symbols TXLNGY
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 60-86 amino acids from the Central region of human CYorf15B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.