Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP11A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597646
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597646 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597646 Supplier Invitrogen™ Supplier No. PA597646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP11A1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CYP11A1, also known as cytochrome P450C11A1, cytochrome P450scc and cytochrome P450, subfamily XIA, is an enzyme that catalyzes the first step of steroid biosynthesis under the modulation of cAMP signal. CYP11A1 in steroidogenic cells converts cholesterol to pregnenolone, which is determined by hormonal control of cholesterol availability. Expression of the CYP11A1 gene is controlled by the transcription factor SF-1, and the upstream SF-1 binding site in the CYP11A1 gene is required for hormonal stimulation. c-Jun and SF-1 may act synergistically to activate CYP11A1 gene expression. Both Forskolin and 8-Br-cAMPS elevate CYP11A1 mRNA levels in the interstitial cell monolayer, which has a fully functional adenylate cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP11A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CYP11A1
Gene Accession No. P05108, P14137
Gene Alias cholesterol 20-22 desmolase; cholesterol desmolase; cholesterol monooxygenase (side-chain cleaving); cholesterol side chain cleavage; cholesterol side-chain cleavage cytochrome P450; cholesterol side-chain cleavage enzyme, mitochondrial; cscc; Cyp11a; Cyp11a1; Cyp11a-1; CYPXIA1; cytochrome P450 11a cholesterol side chain cleavage; cytochrome P450 11A1; cytochrome P450 family 11 subfamily A member 1; cytochrome P450 family 11 subfamily A polypeptide 1; cytochrome P450 side-chain cleavage enzyme; cytochrome P450 subfamily 11A; cytochrome P450(scc); cytochrome P450, 11a, cholesterol side-chain cleavage; cytochrome P450, family 11, subfamily A, polypeptide 1; cytochrome P450, subfamily XIA (cholesterol side chain cleavage); cytochrome P450C11A1; D9Ertd411e; P450(scc); P450scc; Scc; steroid 20-22-lyase
Gene Symbols CYP11A1
Host Species Rabbit
Immunogen Recombinant Human Cholesterol side-chain cleavage enzyme, mitochondrial protein (40-521AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1583, 29680
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.