Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP11B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545063
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545063 Supplier Invitrogen™ Supplier No. PA545063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ARNPNVQQAL RQESLAAAAS ISEHPQKATT ELPLLRAALK ETLRLYPVGL Sequence homology: Human: 100%; Pig: 79%; Sheep: 79%.

The steroid 11beta-hydroxylase gene, also designated Cyp11b-1, is a marker for the functional differentiation of cells in the zonae fasciculata reticularis. The deduced protein CYP11B1 consists of 466 amino acids containing a secretory signal, epidermal growth factor-like repeats, and a proteolytically inactive cathepsin B-related sequence. A related protein, human aldosterone synthase (CYP11B2), is involved in substrate recognition and conversion, with a functionally significant residue 112 in the N-terminal region of human CYP11B2. The inherited disorder glucocorticoid-remediable aldosteronism is caused by a chimeric gene duplication between the CYP11B1 and CYP11B2 genes. This disorder is characterized by hyperaldosteronism and high levels of 18-hydroxycortisol and 18-oxocortisol, which are under ACTH control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP11B1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CYP11B1
Gene Accession No. P15538
Gene Alias 11-beta-hydroxylase; AA389061; aldosterone synthase; Cp11ba; CPN1; CYP11B; CYP11B1; Cyp11b-1; CYPXIB1; Cytochrome P450 11B1, mitochondrial; cytochrome P450 family 11 subfamily B member 1; cytochrome p450 XIB1; cytochrome P450(11 beta)-DS; cytochrome P450, family 11, subfamily b, polypeptide 1; cytochrome P450, subfamily 11B, polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; Cytochrome P450, subfamily XIB, polypeptide 1 (steroid 11-beta-hydroxylase); cytochrome P450C11; cytochrome P-450c11; DKFZp686B05283; FHI; FLJ36771; P450(11 beta)-DS; P450C 11; P450C11; RATCP11BA; S11BH; steroid 11-beta-hydroxylase; steroid 11-beta-hydroxylase, CYP11B1; steroid 11-beta-monooxygenase; steroid-11-beta-hydroxylase
Gene Symbols CYP11B1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CYP11B1 (aa 331-380).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1584
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.