Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP11B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563290 Supplier Invitrogen™ Supplier No. PA563290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RFLPMVDAVA RDFSQALKKK VLQNARGSLT LDVQPSIFHY TIEASNLALF GERLGLVGHS PSSASLNFLH ALEVMFKSTV QLMFMPR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.

The steroid 11beta-hydroxylase gene, also designated Cyp11b-1, is a marker for the functional differentiation of cells in the zonae fasciculata reticularis. The deduced protein CYP11B1 consists of 466 amino acids containing a secretory signal, epidermal growth factor-like repeats, and a proteolytically inactive cathepsin B-related sequence. A related protein, human aldosterone synthase (CYP11B2), is involved in substrate recognition and conversion, with a functionally significant residue 112 in the N-terminal region of human CYP11B2. The inherited disorder glucocorticoid-remediable aldosteronism is caused by a chimeric gene duplication between the CYP11B1 and CYP11B2 genes. This disorder is characterized by hyperaldosteronism and high levels of 18-hydroxycortisol and 18-oxocortisol, which are under ACTH control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP11B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP11B1
Gene Accession No. P15538
Gene Alias 11-beta-hydroxylase; AA389061; aldosterone synthase; Cp11ba; CPN1; CYP11B; CYP11B1; Cyp11b-1; CYPXIB1; Cytochrome P450 11B1, mitochondrial; cytochrome P450 family 11 subfamily B member 1; cytochrome p450 XIB1; cytochrome P450(11 beta)-DS; cytochrome P450, family 11, subfamily b, polypeptide 1; cytochrome P450, subfamily 11B, polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; Cytochrome P450, subfamily XIB, polypeptide 1 (steroid 11-beta-hydroxylase); cytochrome P450C11; cytochrome P-450c11; DKFZp686B05283; FHI; FLJ36771; P450(11 beta)-DS; P450C 11; P450C11; RATCP11BA; S11BH; steroid 11-beta-hydroxylase; steroid 11-beta-hydroxylase, CYP11B1; steroid 11-beta-monooxygenase; steroid-11-beta-hydroxylase
Gene Symbols CYP11B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYP11B1. Recombinant protein control fragment (Product #RP-104168).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1584
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.