Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP11B2 Polyclonal Antibody, Bioss
SDP

Catalog No. 502281829
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1829 Supplier Bioss Supplier No. BS23822R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYP11B2 Polyclonal antibody specifically detects CYP11B2 in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CYP11B2
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene CYP11B2
Gene Accession No. P19099
Gene Alias ALDOS; Aldosterone synthase; aldosterone-synthesizing enzyme; Corticosterone 18-monooxygenase, CYP11B2; Cp45as; Cpn2; Cyp11b; CYP11B2; Cyp11b-2; Cyp11b3; CYP11BL; CYPXIB2; CYPXIB3; cytochrome P450 11B2, mitochondrial; cytochrome P450 11B3, mitochondrial; cytochrome P450 family 11 subfamily B member 2; Cytochrome P450 subfamily XIB polypeptide 2 (aldosterone synthase); cytochrome P450, family 11, subfamily b, polypeptide 2; cytochrome P450, subfamily 11B, polypeptide 2; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2; Cytochrome P450, subfamily XIB, polypeptide 2 (aldosterone synthase); cytochrome P-450Aldo; cytochrome P450-Aldo-1; cytochrome P450-Aldo-2; cytochrome P450C11; Cytochrome P-450C18; mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2; P450aldo; P450-Aldo-1; P450-Aldo-2; P450C 18; P-450C 18; P450C18; P-450C18; RNCP45AS; steroid 11-beta/18-hydroxylase; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B2; steroid 11-be
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CYP11B2
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1585
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.