Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP17A1 Monoclonal Antibody (OTI3F11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525673

Catalog No. PIMA525673


Only null left
Add to Cart

Description

Description

CYP17A1 Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. It has both 17alpha-hydroxylase and 17,20-lyase activities and is a key enzyme in the steroidogenic pathway that produces progestins, mineralocorticoids, glucocorticoids, androgens, and estrogens. Mutations in this gene are associated with isolated steroid-17 alpha-hydroxylase deficiency, 17-alpha-hydroxylase/17,20-lyase deficiency, pseudohermaphroditism, and adrenal hyperplasia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CYP17A1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P05093
CYP17A1
Full length human recombinant protein of CYP17A1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3F11
Unconjugated
CYP17A1
17a-hydroxylase; 17-alpha-hydroxyprogesterone aldolase; CPT7; Cyp17; Cyp17a1; CYPXVII; cytochrome P450 17A1; cytochrome P450 family 17 subfamily A member 1; cytochrome p450 XVIIA1; cytochrome P450, 17; cytochrome P450, 17a1; cytochrome P450, family 17, subfamily A, polypeptide 1; cytochrome P450, subfamily 17; Cytochrome P450, subfamily XVII; cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenal hyperplasia; Cytochrome P450c17; cytochrome P450-C17; HGNC:2593; P45017alpha; p450c17; P450-C17; S17AH; steroid 17-alpha hydroxylase; steroid 17-alpha-hydroxylase/17,20 lyase; Steroid 17-alpha-monooxygenase
Mouse
Affinity Chromatography
RUO
1586
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.