Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP1A1/CYP1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599449 Supplier Invitrogen™ Supplier No. PA599449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 1A1/2.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein encoded by this gene localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. Other xenobiotic substrates for this enzyme include caffeine, aflatoxin B1, and acetaminophen. The transcript from this gene contains four Alu sequences flanked by direct repeats in the 3' untranslated region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP1A1/CYP1A2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP1A2
Gene Accession No. P00184, P00185, P00186, P04798, P04799, P05177
Gene Alias AHH; AHRR; aromatic compound inducible; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP11; CP12; CYP1; Cyp1a1; Cyp1a-1; CYP1A2; Cyp1a-2; Cyp45c; Cypc45c; CYPD45; CYPIA1; CYPIA2; Cytochrome P(3)450; cytochrome P1-450; cytochrome P1-450, dioxin-inducible; Cytochrome P3 450; cytochrome P-448; cytochrome P450 1A1; Cytochrome P450 1A2; cytochrome P450 4; cytochrome P450 family 1 subfamily A member 1; cytochrome P450 family 1 subfamily A member 2; cytochrome P450 family 1 subfamily a polypeptide 1; cytochrome P450 family 1 subfamily a polypeptide 2; cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450 subfamily I, polypeptide 1; cytochrome P450, 1a1; cytochrome P450, 1a1, aromatic compound inducible; cytochrome P450, 1a2; cytochrome P450, 1a2, aromatic compound inducible; cytochrome P450, family 1, subfamily a, polypeptide 1; cytochrome P450, family 1, subfamily a, polypeptide 2; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2; cytochrome P450-C; Cytochrome P-450d; Cytochrome P450-D; Cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; cytochrome P450-P1; Cytochrome P450-P2; cytochrome P450-P3; dioxin-inducible; dioxin-inducible P3-450; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P1-450; P3-450; P-448; P450 form 4; P450(PA); P450-1; P450-3; P450-C; P-450d; P450-D; P450DX; P450form6; P-450MC; P450MT2; RATCYPD45; xenobiotic monooxygenase
Gene Symbols CYP1A1, CYP1A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP1A1(Accession P04798), corresponding to amino acid residues I78-G128.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13076, 13077, 1543, 1544, 24296, 24297
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.