Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP1A2 Monoclonal Antibody (OTI7D12), TrueMAB™, OriGene
SDP

Catalog No. 502542480
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2480 30 μL
50-167-2317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2480 Supplier OriGene Supplier No. TA501173S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP1A2 Monoclonal antibody specifically detects CYP1A2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CYP1A2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7D12
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Cyp1a2
Gene Accession No. P05177
Gene Alias aromatic compound inducible; aryl hydrocarbon hydroxylase; cholesterol 25-hydroxylase; CP12; CYP; Cyp1a1; Cyp1a2; Cyp1a-2; CYPD45; CYPIA2; Cytochrome P(3)450; cytochrome P-448; cytochrome P450 1A2; Cytochrome P450 4; cytochrome P450 family 1 subfamily A member 2; cytochrome P450 family 1 subfamily a polypeptide 1; Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450, 1a2; cytochrome P450, 1a2, aromatic compound inducible; cytochrome P450, family 1, subfamily A, polypeptide 2; cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 2; Cytochrome P-450d; cytochrome P450-D; Cytochrome P450-P2; cytochrome P450-P3; dioxin-inducible P3-450; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P(3)450; P3-450; P-448; P450; P450 4; P450 form 4; P450(PA); P450-3; P-450d; P450-D; P450-P3; RATCYPD45; xenobiotic monooxygenase
Host Species Mouse
Immunogen Full length human recombinant protein of human CYP1A2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1544
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.