Learn More
CYP1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 185051AP
Catalog No. 50-173-2171
Add to cart
Description
Description
CYP1B1 is a 543 amino acid intracellular protein localized to the endoplasmic reticulum. It belongs to the cytochrome P450 super family of enzymes which are monooxygenases and it catalyzes many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme metabolizes procarcinogens such as polycyclic aromatic hydrocarbons and 17beta-estradiol and is also involved in the metabolism of substances active in the eye growth and differentiation and hence plays a putative role in ocular development. Mutated proteins are known to cause primary congenital glaucoma.
Specifications
Specifications
CYP1B1
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16678, Q64429, Q64678
CP1B, CYP1B1, CYPIB1, Cytochrome P450 1B1, GLC3A, P4501B1
Rabbit
IgG
150 μL
-20°C
Primary
13078, 1545, 25426
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CYP1B1
Liquid
Cyp1b1
CYP1B1 Fusion Protein Ag13357
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat