Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP20A1 Antibody (Z27-P3H7*E3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250192AF594
Description
CYP20A1 Monoclonal specifically detects CYP20A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP20A1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CYP20A1 | |
Ovalbumin-conjugated synthetic peptide AWITVSKRY | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z27-P3H7*E3 | |
Western Blot, ELISA, Immunohistochemistry | |
CYP-M, cytochrome P450 20A1, cytochrome P450 monooxygenase, cytochrome P450, family 20, subfamily A, polypeptide 1, EC 1.14, MGC22229 | |
Mouse | |
Protein A purified | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
57404.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction