Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP20A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102020
Description
Antibody detects endogenous levels of total Cytochrome P450 20A1.
CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.
Specifications
CYP20A1 | |
Polyclonal | |
Unconjugated | |
CYP20A1 | |
A930011N14Rik; Cyp20a1; Cypm; CYP-M; cytochrome P450 20A1; cytochrome P450 family 20 subfamily A member 1; cytochrome P450 monooxygenase; cytochrome P450, 20a1; cytochrome P450, family 20, subfamily A, polypeptide 1; UNQ667/PRO1301 | |
Rabbit | |
Affinity chromatography | |
RUO | |
57404 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6UW02 | |
CYP20A1 | |
A synthesized peptide derived from human CYP20A1(Accession Q6UW02), corresponding to amino acid residues F356-T406. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction