Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP20A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109611 Supplier Invitrogen™ Supplier No. PA5109611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STFEDDQEVI RFQKNHGTVW SEIGKGFLDG SLDKNMTRKK QYEDALMQLE SVLRNIIKER KGRNFSQHIF IDSLVQGNLN DQQILEDSMI FSLASCIITA KLCTWAICFL TTSEEVQKKL YEEINQVFGN GPVTPEKIEQ LRYCQHVLCE TVRTAKLTPV SAQLQDIEGK IDRFIIPRET LVLYALGVVL QDPNTWPSPH KFDPDRFDDE LVMKTFSSLG FSGTQECPEL RFAYMVTTVL LSVLVKRLHL LSVEGQVIET KYELVTSSRE EAWITVSKRY.

CYP20A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. The protein lacks one amino acid of the conserved heme binding site. It also lacks the conserved I-helix motif AGX(D, E)T, suggesting that its substrate may carry its own oxygen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP20A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP20A1
Gene Accession No. Q6P7D4, Q6UW02, Q8BKE6
Gene Alias A930011N14Rik; Cyp20a1; Cypm; CYP-M; cytochrome P450 20A1; cytochrome P450 family 20 subfamily A member 1; cytochrome P450 monooxygenase; cytochrome P450, 20a1; cytochrome P450, family 20, subfamily A, polypeptide 1; UNQ667/PRO1301
Gene Symbols CYP20A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 183-462 of human CYP20A1 (NP_803882.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316435, 57404, 77951
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.