Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP21A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529441 Supplier Invitrogen™ Supplier No. PA529441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4. Predicted reactivity: Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CYP21A2 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and hydroxylates steroids at the 21 position. Its activity is required for the synthesis of steroid hormones including cortisol and aldosterone. Mutations in CYP21A2 gene cause congenital adrenal hyperplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP21A2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CYP21A2
Gene Accession No. P08686
Gene Alias 21-OHase; CA21H; CAH1; CPS1; CYP21; CYP21A2; CYP21B; Cytochrome P450 21; cytochrome P450 family 21 subfamily A member 2; cytochrome P450 XXI; cytochrome P450, family 21, subfamily A, polypeptide 2; cytochrome P450, subfamily XXIA (steroid 21-hydroxylase, congenital adrenal hyperplasia), polypeptide 2; Cytochrome P-450c21; Cytochrome P450-C21; Cytochrome P450-C21B; P-450c21; P450-C21; P450c21B; P450-C21B; steroid 21 hydroxylase; steroid 21-hydroxylase; steroid 21-monooxygenase
Gene Symbols CYP21A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CYP21A2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.