Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP24A1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305981AF594
Description
CYP24A1 Polyclonal specifically detects CYP24A1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CYP24A1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial, CP241, CYP241,25-alphadihydroxyvitamin D3 24-hydroxylase, Cytochrome P450 24A1, cytochrome P450, family 24, subfamily A, polypeptide 1,24-OHase, cytochrome P450, subfamily XXIV (vitamin D 24-hydroxylase), Cytochrome P450-CC24, EC 1.14.13.n4, exo-mitochondrial protein, MGC126273, MGC126274, P450-CC24, vitamin D 24-hydroxylase, Vitamin D(3) 24-hydroxylase | |
Produced in rabbits immunized with E. coli-derived Human CYP24A1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
1591 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction