Learn More
CYP26A1 Antibody (F27P6A1), Janelia Fluor™ 549, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP250193JF549
Catalog No. NBP50193549
Only null left
Add to Cart
Description
Description
CYP26A1 Monoclonal specifically detects CYP26A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
CYP26A1
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CYP26A1
Ovalbumin-conjugated synthetic peptide PARFTHFHGE (C-terminal sequence)
0.1 mL
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
F27P6A1
Western Blot, ELISA, Immunohistochemistry
CP26, CYP26Retinoic acid 4-hydroxylase, cytochrome P450 26A1, Cytochrome P450 retinoic acid-inactivating 1, cytochrome P450, family 26, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIA, polypeptide 1, Cytochrome P450RAI, EC 1.14, hP450RAI, P450, retinoic acid-inactivating, 1, P450RAI, P450RAI1Retinoic acid-metabolizing cytochrome
Mouse
Protein A purified
Stem Cell Markers
1592
Store at 4°C in the dark.
IgG2b κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.