Learn More
Invitrogen™ CYP26B1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total CYP26B1.
Specifications
Specifications
| Antigen | CYP26B1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CYP26B1 |
| Gene Accession No. | G3V7X8, Q811W2, Q9NR63 |
| Gene Alias | CP26; CYP26A2; CYP26B1; Cytochrome P450 26A2; Cytochrome P450 26B1; cytochrome P450 family 26 subfamily A member 1; cytochrome P450 family 26 subfamily B member 1; cytochrome P450 retinoic acid-inactivating 2; cytochrome P450 retinoid metabolizing protein; cytochrome P450, 26, retinoic acid B1; cytochrome P450, family 26, subfamily b, polypeptide 1; cytochrome P450, subfamily XXVIB, polypeptide 1; cytochrome P450RAI-2; dol; fc21d03; P450 26A2; P450 enzyme to degrade retinoic acid; P450RAI2; P450RAI-2; retinoic acid B1; retinoic acid hydroxylase; retinoic acid-metabolizing cytochrome; RHFCA; stocksteif; wu:fc21d03; wu:fc26h10; zgc:76999 |
| Gene Symbols | CYP26B1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.