Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP26C1 Antibody (T6P1C7*E7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250196G
Description
CYP26C1 Monoclonal specifically detects CYP26C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CYP26C1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP26C1 | |
Ovalbumin-conjugated synthetic peptide RWELATPAFP (C-terminal sequence) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
T6P1C7*E7 | |
Western Blot, Immunohistochemistry | |
cytochrome P450 26C1, cytochrome P450, family 26, subfamily C, polypeptide 1, EC 1.14, FLJ45301 | |
Mouse | |
Protein A purified | |
RUO | |
340665 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction