Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP26C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144609 Supplier Invitrogen™ Supplier No. PA5144609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP26C1 Polyclonal Antibody for Western Blot

CYP26C1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is involved in the catabolism of all-trans- and 9-cis-retinoic acid, and thus contributes to the regulation of retinoic acid levels in cells and tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP26C1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal
Gene CYP26C1
Gene Alias CYP26C1; Cytochrome P450 26C1; cytochrome P450 family 26 subfamily C member 1; cytochrome P450, family 26, subfamily c, polypeptide 1; EG546726; FFDD4; predicted gene, EG546726
Gene Symbols CYP26C1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of zebrafish Cyp26c1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 554036
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.