Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP26C1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575859
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
CYP26C1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is involved in the catabolism of all-trans- and 9-cis-retinoic acid, and thus contributes to the regulation of retinoic acid levels in cells and tissues.
Specifications
CYP26C1 | |
Polyclonal | |
Unconjugated | |
CYP26C1 | |
CYP26C1; Cytochrome P450 26C1; cytochrome P450 family 26 subfamily C member 1; cytochrome P450, family 26, subfamily c, polypeptide 1; EG546726; FFDD4; predicted gene, EG546726 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
308190, 340665, 546726 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6V0L0 | |
CYP26C1 | |
Synthetic peptide corresponding to residues in Human CYP26C1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction