Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP27A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596021 Supplier Invitrogen™ Supplier No. PA596021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NQVSDMAQLF YYFALEAICY ILFEKRIGCL QRSIPEDTVT FVRSIGLMFQ NSLYATFLPK WTRPVLPFWK RYLDGWNAIF SFGKKLIDEK LEDMEAQLQA AGPDGIQVSG YLHFLLASGQ LSPREAMGSL PELLMAGVDT TSNTLTWALY HLSKDPEIQE ALHEEVVGVV PAGQVPQHKD FAHMPLLKAV LKETLRLYPV V; Positive Samples: 293T, Mouse kidney, Rat liver; Cellular Location: Mitochondrion membrane.

CYP27A1 participates in the degradation of cholesterol to bile acids in both the classic and acidic pathways. [1] It is the initiating enzyme in the acidic pathway to bile acids, yielding oxysterols by introducing a hydroxyl group to the carbon at the 27 position in cholesterol. In the acidic pathway, it produces 27-hydroxycholesterol from cholesterol whereas in the classic or neutral pathway, it produces 3beta-hydroxy-5-cholestenoic acid. It is also involved in the metabolism of vitamin D3. While CYP27A1 is present in many different tissues, its function in these tissues is largely uncharacterized. In macrophages, 27-hydroxycholesterol generated by this enzyme may be helpful against the production of inflammatory factors associated with cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP27A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CYP27A1
Gene Accession No. P17178, Q02318, Q9DBG1
Gene Alias 1300013A03Rik; 5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 26-hydroxylase; 5-beta-cholestane-3-alpha, 7-alpha, 12-alpha-triol 27-hydroxylase; 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 26-hydroxylase; 5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase; cholestanetriol 26-monooxygenase; cholesterol 27 hydroxylase; CP27; CTX; CYP27; CYP27A1; cyt; cytochrome P450 27; cytochrome P450 family 27 subfamily A member 1; cytochrome P450, 27; cytochrome P450, 27a1; cytochrome P450, family 27, subfamily a, polypeptide 1; cytochrome P450, subfamily XXVIIA (steroid 27-hydroxylase, cerebrotendinous xanthomatosis), polypeptide 1; Cytochrome P-450C27/25; P450C27; Sterol 26-hydroxylase, mitochondrial; sterol 27-hydroxylase; Vitamin D(3) 25-hydroxylase
Gene Symbols CYP27A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human CYP27A1 (NP_0007751).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104086, 1593, 301517
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.