Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP27C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584549 Supplier Invitrogen™ Supplier No. PA584549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTSFTLSWTV YLLARHPEVQ QTVYREIVKN LGERHVPTAA DVPKVPLVRA LLK Highest antigen sequence indentity to the following orthologs: Mouse - 43%, Rat - 42%.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP27C1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP27C1
Gene Accession No. Q4G0S4
Gene Alias All-trans retinol 3,4-desaturase; CYP27C1; Cytochrome P450 27C1; cytochrome P450 family 27 subfamily C member 1; cytochrome P450, family 27, subfamily C, polypeptide 1; cytochrome P450, family 27, subfamily C, polypeptide 13
Gene Symbols CYP27C1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYP27C1. Recombinant protein control fragment (Product #RP-108471).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339761
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.