Learn More
Invitrogen™ CYP2A6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA1342
Description
PA1-342 detects cytochrome P450 (P450) 2A6 from human, mouse and rat samples. This antibody detects, to a lesser extent, other P450 isoforms. PA1-342 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~49 kDa protein representing P450 2A6 from rat liver extract. The PA1-342 immunogen is purified, His-tagged, full length human P450 2A6 fusion protein.
This gene, CYP2A6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to hydroxylate coumarin, and also metabolizes nicotine, aflatoxin B1, nitrosamines, and some pharmaceuticals. Individuals with certain allelic variants are said to have a poor metabolizer phenotype, meaning they do not efficiently metabolize coumarin or nicotine. This gene is part of a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q. The gene was formerly referred to as CYP2A3; however, it has been renamed CYP2A6.
Specifications
CYP2A6 | |
Polyclonal | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P11509 | |
Cyp2a, CYP2A6 | |
Purified, His-tagged, full length human P450 2A6 fusion protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CYP2A6 | |
1,4-cineole 2-exo-monooxygenase; Coumarin 7-hydroxylase; CPA6; CYP; CYP2A; CYP2A3; CYP2A6; CYPIIA6; cytochrome P450 2A6; cytochrome P450 family 2 subfamily A member 6; cytochrome P450 IIA3; Cytochrome P450(I); cytochrome P450, family 2, subfamily A, polypeptide 6; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6; flavoprotein-linked monooxygenase; P450; P450 IIA3; P450(I); P450C2A; P450PB; xenobiotic monooxygenase | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
110690, 1548 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.