Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP2A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527890
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene, CYP2A6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to hydroxylate coumarin, and also metabolizes nicotine, aflatoxin B1, nitrosamines, and some pharmaceuticals. Individuals with certain allelic variants are said to have a poor metabolizer phenotype, meaning they do not efficiently metabolize coumarin or nicotine. This gene is part of a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q. The gene was formerly referred to as CYP2A3; however, it has been renamed CYP2A6.
Specifications
| CYP2A6 | |
| Polyclonal | |
| Unconjugated | |
| CYP2A6 | |
| 1,4-cineole 2-exo-monooxygenase; Coumarin 7-hydroxylase; CPA6; CYP; CYP2A; CYP2A3; CYP2A6; CYPIIA6; cytochrome P450 2A6; cytochrome P450 family 2 subfamily A member 6; cytochrome P450 IIA3; Cytochrome P450(I); cytochrome P450, family 2, subfamily A, polypeptide 6; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6; flavoprotein-linked monooxygenase; P450; P450 IIA3; P450(I); P450C2A; P450PB; xenobiotic monooxygenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| P11509 | |
| CYP2A6 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Cytochrome P450 2A6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction