Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP2A6/CYP2A7/CYP2A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599833
Description
Antibody detects endogenous levels of total Cytochrome P450 2A6/7/13.
The cytochrome P450 (P450) superfamily of enzymes is one of three enzyme systems which metabolize the fatty acid arachadonic acid (AA) to regulators of vascular tone. P450 enzymes are monooxygenase enzymes which require several co-factors such as nicotinamide adenine dinucleotide phosphate (NADPH) and P450 reductase. There are over 200 known genes which encode P450s. Epoxygenases are those P450s which metabolize AA to epoxyeicosatrienoic acid (EETs) and omega-hydroxylases are those P450s which produce 19- and 20-hydroxyeicosatetraenoic acids (19- and 20-HETE). As well as fatty acid metabolism, P450s also metabolize many drugs and toxins. Cytochrome P450 3A4 is abundantly expressed in liver and small intestine and is inducible by barbiturates, glucocorticoids and rifampicin.
Specifications
CYP2A6/CYP2A7/CYP2A13 | |
Polyclonal | |
Unconjugated | |
CYP2A6 | |
1,4-cineole 2-exo-monooxygenase; Coumarin 7-hydroxylase; CPA6; CPA7; CPAD; CYP2A; CYP2A13; CYP2A3; CYP2A6; CYP2A7; CYPIIA13; CYPIIA6; CYPIIA7; Cytochrome P450 2A13; cytochrome P450 2A6; Cytochrome P450 2A7; cytochrome P450 family 2 subfamily A member 13; cytochrome P450 family 2 subfamily A member 6; cytochrome P450 family 2 subfamily A member 7; cytochrome P450 IIA3; Cytochrome P450 IIA4; cytochrome P450(I); cytochrome P450, family 2, subfamily A, polypeptide 13; cytochrome P450, family 2, subfamily A, polypeptide 6; cytochrome P450, family 2, subfamily A, polypeptide 7; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 13; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6; cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 7; cytochrome P450IIA4; flavoprotein-linked monooxygenase; P450C2A; P450-IIA4; P450PB; xenobiotic monooxygenase | |
Rabbit | |
Affinity chromatography | |
RUO | |
1548, 1549, 1553 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P11509, P20853, Q16696 | |
CYP2A13, CYP2A6, CYP2A7 | |
A synthesized peptide derived from human CYP2A6(Accession P11509), corresponding to amino acid residues S4-M54. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction