Learn More
Invitrogen™ CYP2B6 Polyclonal Antibody

Description
Immunogen sequence: EAQCLIEELRK SKGALVDPTF LFHSITANII CSI Highest antigen sequence identity to the following orthologs - mouse 82%, rat 82%.
Specifications
Specifications
| Antigen | CYP2B6 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CYP2B6 |
| Gene Accession No. | P20813 |
| Gene Alias | 1,4-cineole 2-exo-monooxygenase; CPB6; CYP2B; CYP2B11; Cyp2b3; Cyp2b-3; CYP2B30; CYP2B6; CYP2B7; CYP2B7P; CYPIIB11; CYPIIB3; CYPIIB6; cytochrome P450 2B11; Cytochrome P450 2B3; cytochrome P450 2B6; cytochrome P450 CYP2B30; cytochrome P450 family 2 subfamily B member 6; cytochrome P-450 IIB; cytochrome P450 IIB1; Cytochrome P450 PBD-2; cytochrome P450, family 2, subfamily B, polypeptide 11; cytochrome P450, family 2, subfamily b, polypeptide 3; cytochrome P450, family 2, subfamily B, polypeptide 6; cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6; cytochrome P450IIB3; Cytochrome-P450-2B11; EFVM; IIB1; P450 |
| Gene Symbols | CYP2B6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.