Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2B6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535032
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535032 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535032 Supplier Invitrogen™ Supplier No. PA535032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP2B6 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

This gene, CYP2B6, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to metabolize some xenobiotics, such as the anti-cancer drugs cyclophosphamide and ifosphamide. Transcript variants for this gene have been described; however, it has not been resolved whether these transcripts are in fact produced by this gene or by a closely related pseudogene, CYP2B7. Both the gene and the pseudogene are located in the middle of a CYP2A pseudogene found in a large cluster of cytochrome P450 genes from the CYP2A, CYP2B and CYP2F subfamilies on chromosome 19q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2B6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CYP2B6
Gene Accession No. P20813
Gene Alias 1,4-cineole 2-exo-monooxygenase; CPB6; CYP2B; CYP2B11; Cyp2b3; Cyp2b-3; CYP2B30; CYP2B6; CYP2B7; CYP2B7P; CYPIIB11; CYPIIB3; CYPIIB6; cytochrome P450 2B11; Cytochrome P450 2B3; cytochrome P450 2B6; cytochrome P450 CYP2B30; cytochrome P450 family 2 subfamily B member 6; cytochrome P-450 IIB; cytochrome P450 IIB1; Cytochrome P450 PBD-2; cytochrome P450, family 2, subfamily B, polypeptide 11; cytochrome P450, family 2, subfamily b, polypeptide 3; cytochrome P450, family 2, subfamily B, polypeptide 6; cytochrome P450, subfamily IIB (phenobarbital-inducible), polypeptide 6; cytochrome P450IIB3; Cytochrome-P450-2B11; EFVM; IIB1; P450
Gene Symbols CYP2B6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 235-263 amino acids from the central region of human CYP2B6.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1555
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.