Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2C18 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989780
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9780 Supplier Bioss Supplier No. BS14150R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CYP2C18
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene CYP2C18
Gene Accession No. P33260
Gene Alias (S)-mephenytoin hydroxylase associated cytochrome P450; CPCI; CYP2C; CYP2C17; CYP2C18; CYPIIC18; Cytochrome P450 2C18; cytochrome P450 family 2 subfamily C member 18; cytochrome P450, family 2, subfamily C, polypeptide 18; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18; Cytochrome P450-6b/29c; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450-6B/29C; P450IIC17; unspecific monooxygenase
Gene Symbols CYP2C18
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CYP2C18.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1562
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.