Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2C19 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501736168 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6168 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6168 Supplier Proteintech Group Inc Supplier No. 554531AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CYP2C19 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, omeprazole, diazepam and some barbiturates. Polymorphism within its gene is associated with variable ability to metabolize mephenytoin, known as the poor metabolizer and extensive metabolizer phenotypes.

Specifications

Antigen CYP2C19
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYP2C19
Gene Accession No. P33261
Gene Alias (R) limonene 6 monooxygenase, (S) limonene 6 monooxygenase, (S) limonene 7 monooxygenase, CPCJ, CYP2C, CYP2C19, CYPIIC17, CYPIIC19, Cytochrome P450 11A, Cytochrome P450 254C, Cytochrome P450 2C19, Mephenytoin 4 hydroxylase, P450C2C, P450IIC19
Gene Symbols CYP2C19
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1557
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.