Learn More
Invitrogen™ CYP2C8 Polyclonal Antibody

Description
Immunogen sequence: ICSVVFQKRF DYKDQNFLTL MKRFNENFRI LNSPWIQVCN NFPLLIDCFP GTHNKVLKNV ALTRSYIREK VKEHQASLDV NNPRDFIDCF LIKMEQEKDN QKSEFNIENL VGTVADLFVA G Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 68%.
Specifications
Specifications
| Antigen | CYP2C8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CYP2C8 |
| Gene Accession No. | P10632 |
| Gene Alias | CPC8; CYP2C8; CYPIIC8; Cytochrome P450 2C8; cytochrome P450 family 2 subfamily C member 8; cytochrome P450 form 1; Cytochrome P450 IIC2; cytochrome P450 MP-12; Cytochrome P450 MP-20; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase; MP-12/MP-20; P450 form 1; P450 IIC2; P450 MP-12/MP-20; s-mephenytoin 4-hydroxylase; xenobiotic monooxygenase |
| Gene Symbols | CYP2C8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.