Learn More
Description
Specifications
Specifications
| Antigen | CYP2C8 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CYP2C8 |
| Gene Accession No. | P10632 |
| Gene Alias | CPC8, CYP2C8, CYPIIC8, Cytochrome P450 2C8, Cytochrome P450 form 1, Cytochrome P450 IIC2, Cytochrome P450 MP 12, Cytochrome P450 MP 20, MP 12/MP 20, S mephenytoin 4 hydroxylase |
| Gene Symbols | CYP2C8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
