Learn More
Invitrogen™ CYP2C9 Monoclonal Antibody (OTI1D7)

Description
CYP2C9 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow
Specifications
Specifications
| Antigen | CYP2C9 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1D7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CYP2C9 |
| Gene Accession No. | P11712 |
| Gene Alias | (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; Cholesterol 25-hydroxylase; CPC9; CYP2C; CYP2C10; CYP2C9; CYPIIC9; Cytochrome P450 2C9; cytochrome P450 family 2 subfamily C member 9; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P-450 S-mephenytoin 4-hydroxylase; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P-450MP; flavoprotein-linked monooxygenase; MGC149605; MGC88320; microsomal monooxygenase; P450IIC9; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.