Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2D10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537998
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537998 Supplier Invitrogen™ Supplier No. PA537998
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2D10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Cyp2d5
Gene Accession No. P12939
Gene Alias Cyp2d10; Cyp2d-10; Cyp2d5; CYPIID10; cytochrome P450 2D10; cytochrome P450, family 2, subfamily d, polypeptide 10; cytochrome P450, family 2, subfamily d, polypeptide 5; cytochrome P450CMF1b; cytochrome P450-CMF1B; Cytochrome P450-DB5; debrisoquine 4-hydroxylase; MGC108540; P450-CMF1B; P450-DB5
Gene Symbols Cyp2d5
Host Species Goat
Immunogen Peptide with sequence CDAFTAQNGRSINPK, from the internal region of rat Cyp2d5.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286963
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.