Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP2D6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586925
Description
Purity is > 95% by SDS-PAGE.
Cytochrome p450 2D6 (CYP2D6) is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant. The gene is highly polymorphic in the population; certain alleles result in the poor metabolizer phenotype, characterized by a decreased ability to metabolize the enzyme's substrates. The gene is located near two cytochrome P450 pseudogenes on chromosome 22q13.1. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
CYP2D6 | |
Polyclonal | |
Unconjugated | |
CYP2D6 | |
AI303445; Cholesterol 25-hydroxylase; CPD6; CYP2D; Cyp2d10; Cyp2d-10; Cyp2d13; Cyp2d18; Cyp2d-18; Cyp2d22; Cyp2d3; Cyp2d-3; Cyp2d4; Cyp2d-4; Cyp2d4v1; Cyp2d4v2; Cyp2d6; CYP2D7AP; CYP2D7BP; CYP2D7P2; CYP2D8P2; CYP2DL1; CYPIID10; CYPIID18; CYPIID3; CYPIID4; CYPIID6; Cytochrome P450 2D10; cytochrome P450 2d18; Cytochrome P450 2D-29; Cytochrome P450 2D3; cytochrome P450 2D-35; cytochrome P450 2D4; Cytochrome P450 2D6; cytochrome P450 family 2 subfamily D member 6; cytochrome P450, 2d10; cytochrome P450, family 2, subfamily d, polypeptide 10; cytochrome P450, family 2, subfamily d, polypeptide 13; cytochrome P450, family 2, subfamily d, polypeptide 22; cytochrome P450, family 2, subfamily d, polypeptide 3; cytochrome P450, family 2, subfamily d, polypeptide 4; cytochrome P450, family 2, subfamily d, polypeptide 6; cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2; cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2; cytochrome P450, subfamily 2D, polypeptide 6; cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), polypeptide 7 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), polypeptide 8 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), polypeptide 6; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing)-like 1; cytochrome P450, subfamily IID, polypeptide 6; Cytochrome P450, subfamily IID3; Cytochrome P450, subfamily IID4; cytochrome P450-16-alpha; Cytochrome P450CB; Cytochrome P450-CMF3; cytochrome P450-DB1; Cytochrome P450-DB3; cytochrome P450-DB4; debrisoquine 4-hydroxylase; flavoprotein-linked monooxygenase; microsomal monooxygenase; nonfunctional cytochrome P450 2D6; P450 2D-29/2D-35; P450-2D; P450C2D; P450-CMF3; P450DB1; P450-DB1; P450-DB3; P450-DB4; testosterone 16-alpha hydroxylase; xenobiotic monooxygenase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1565, 171522, 24303 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P10635, P12938, Q64680 | |
Cyp2d3, Cyp2d4, CYP2D6 | |
Synthetic peptide, corresponding to amino acids 250-300 of Human CYP2D6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction