Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2E1 Mouse anti-Human, Unconjugated, Clone: M12P4H2, Abnova™

Catalog No. 89337401 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-401 Supplier Abnova Corporation Supplier No. MAB20906
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CYP2E1 Monoclonal antibody specifically detects CYP2E1 in Human samples. It is validated for IHC-P.

Specifications

Antigen CYP2E1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M12P4H2
Conjugate Unconjugated
Description cytochrome P450, family 2, subfamily E, polypeptide 1
Gene CYP2E1
Gene Alias CPE1, CYP2E, P450-J, P450C2E
Host Species Mouse
Immunogen A synthetic peptide (conjugated with Ovalbumin) corresponding to C-terminus region of human CYP2E1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1571
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.