Learn More
Invitrogen™ CYP2E1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120794
Description
Positive test controls include: Mouse liver, Mouse kidney, Rat liver. The target is usually found in the following locations: Endoplasmic reticulum membrane, Microsome membrane, Peripheral membrane protein. Immunogen sequence: KVIKNVAEVK EYVSERVKEH HQSLDPNCPR DLTDCLLVEM EKEKHSAERL YTMDGITVTV ADLFFAGTET TSTTLRYGLL ILMKYPEIEE KLHEEIDRVI GPSRIPAIKD RQEMPYMDAV VHEIQRFITL VPSNLPHEAT RDTIFRGYLI PKGTVVVPTL DSVLYDNQEF PDPEKFKPEH FLNENGKFKY SDYFKPFSTG KRVCAGEGLA RMELFLLLCA ILQHFNLKPL VDPKDIDLSP IHIGFGCIPP RYKLCVIPRS.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.
Specifications
| CYP2E1 | |
| Polyclonal | |
| Unconjugated | |
| Cyp2e1 | |
| 4-nitrophenol 2-hydroxylase; CP2E1; CPE1; Cyp2e; Cyp2e1; Cyp2e-1; CYPIIE1; cytochrome P450 2E1; cytochrome P450 family 2 subfamily E member 1; cytochrome P450 subfamily 2e1 (ethanol-inducible); cytochrome P450, 2e1, ethanol inducible; cytochrome P450, family 2, subfamily e, polypeptide 1; cytochrome P450, subfamily 2E, polypeptide 1; cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1; cytochrome P450-ALC; Cytochrome P450-J; cytochrome P450RLM6; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450C2E; P450-J; P450RLM6; xenobiotic monooxygenase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13106, 1571, 25086 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P05181, P05182, Q05421 | |
| Cyp2e1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 234-493 of human CYP2E1 (NP_000764.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.