Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CYP2E1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519381
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is induced by ethanol, the diabetic state, and starvation. The enzyme metabolizes both endogenous substrates, such as ethanol, acetone, and acetal, as well as exogenous substrates including benzene, carbon tetrachloride, ethylene glycol, and nitrosamines which are premutagens found in cigarette smoke. Due to its many substrates, this enzyme may be involved in such varied processes as gluconeogenesis, hepatic cirrhosis, diabetes, and cancer.
Specifications
| CYP2E1 | |
| Polyclonal | |
| Unconjugated | |
| Cyp2e1 | |
| 4-nitrophenol 2-hydroxylase; CP2E1; CPE1; Cyp2e; Cyp2e1; Cyp2e-1; CYPIIE1; cytochrome P450 2E1; cytochrome P450 family 2 subfamily E member 1; cytochrome P450 subfamily 2e1 (ethanol-inducible); cytochrome P450, 2e1, ethanol inducible; cytochrome P450, family 2, subfamily e, polypeptide 1; cytochrome P450, subfamily 2E, polypeptide 1; cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1; cytochrome P450-ALC; Cytochrome P450-J; cytochrome P450RLM6; flavoprotein-linked monooxygenase; microsomal monooxygenase; P450C2E; P450-J; P450RLM6; xenobiotic monooxygenase | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1571 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P05181 | |
| Cyp2e1 | |
| Synthetic peptide sequence (RKVIKNVAEVK) corresponding to the internal amino acids of CYP2E1 (aa 233-244). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction