Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2F1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2839860.1ML
Description
CYP2F1 Polyclonal specifically detects CYP2F1 in Human samples. It is validated for Western Blot.Specifications
CYP2F1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
C2F1, CYP2F, CYPIIF1, cytochrome P450 2F1, cytochrome P450, family 2, subfamily F, polypeptide 1, cytochrome P450, subfamily IIF, polypeptide 1, EC 1.14.14.1, flavoprotein-linked monooxygenase, MGC126121, microsomal monooxygenase, xenobiotic monooxygenase | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human CYP2F1 | |
0.1 mL | |
Lipid and Metabolism | |
1572 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction