Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2J2 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
SDP

Catalog No. 502540866
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0866 30 μL
50-167-4387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0866 Supplier OriGene Supplier No. TA503515S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CYP2J2 Monoclonal antibody specifically detects CYP2J2 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen CYP2J2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4B9
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CTBP1-DT
Gene Alias Albendazole monooxygenase (hydroxylating); Albendazole monooxygenase (sulfoxide-forming); Arachidonic acid epoxygenase; CPJ2; CYP2J2; Cyp2j4; CYPIIJ2; cytochrome P450 2J2; cytochrome P450 family 2 subfamily J member 2; cytochrome P450, family 2, subfamily J, polypeptide 2; cytochrome P450, family 2, subfamily j, polypeptide 4; cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate isomerase; microsomal monooxygenase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92070
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.