Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2J2 Mouse anti-Mouse, Human, Clone: 6-5-20-8, MilliporeSigma™

Catalog No. MABS1242MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1242MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1242MI Supplier MilliporeSigma Supplier No. MABS1242
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CYP2J2, clone 6-5-20-8 is a mouse monoclonal antibody that targets cytochrome P450 2J2 and is tested for use in Inhibition Assay and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2J2
Applications Western Blot, Inhibition Assays
Classification Monoclonal
Clone 6-5-20-8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CYP2J2
Host Species Mouse
Immunogen A full-length recombinant human cytochrome P450 2J2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000766
Test Specificity Clone 6-5-20-8 detects cytochrome p450 2J2 in human and murine species.
Target Species Mouse, Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.