Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2R1 Antibody (M26-P6H1), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250204C
Description
CYP2R1 Monoclonal specifically detects CYP2R1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP2R1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP2R1 | |
Ovalbumin-conjugated synthetic peptide QPYLICAERR (C-terminal sequence). | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
120227 | |
Store at 4C in the dark. | |
IgG1 λ |
Western Blot, ELISA, Immunohistochemistry | |
M26-P6H1 | |
Western Blot, ELISA, Immunohistochemistry | |
Cytochrome P450 2R1, cytochrome P450, family 2, R1, cytochrome P450, family 2, subfamily R, polypeptide 1, EC 1.14.13.15, MGC4663, vitamin D 25-hydroxylase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction