Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111167 Supplier Invitrogen™ Supplier No. PA5111167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYEDTDFQHM IELFSENVEL AASASVFLYN AFPWIGILPF GKHQQLFRNA AVVYDFLSRL IEKASVNRKP QLPQHFVDAY LDEMDQGKND PSSTFSKENL IFSVGELIIA GTETTTNVLR Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 92%.

CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor. A mutation in CYP2R1 gene has been associated with selective 25-hydroxyvitamin D deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2R1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cyp2r1
Gene Accession No. Q6VVX0
Gene Alias Cyp2r1; Cytochrome P450 2R1; cytochrome P450 family 2 subfamily R member 1; cytochrome P450, 2r1; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; vitamin D 25-hydroxylase
Gene Symbols Cyp2r1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CYP2R1. Recombinant protein control fragment (Product #RP-90562).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.