Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2U1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179914
Description
CYP2U1 Polyclonal specifically detects CYP2U1 in Human samples. It is validated for Western Blot.Specifications
| CYP2U1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cytochrome P450 2U1, cytochrome P450, family 2, subfamily U, polypeptide 1, EC 1.14.14.1, P450TEC | |
| Rabbit | |
| 69 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 86%; Dog: 79%; Pig: 79%; Rat: 79%; Goat: 79%; Horse: 79%; Sheep: 79%; Mouse: 77%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Goat, Rabbit, Sheep | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_898898 | |
| CYP2U1 | |
| The immunogen for this antibody is CYP2U1. Peptide sequence VNICPWLYYLPFGPFKELRQIEKDITSFLKKIIKDHQESLDRENPQDFID. | |
| Affinity purified | |
| RUO | |
| 113612 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction