Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2U1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559650 Supplier Invitrogen™ Supplier No. PA559650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPFGPFKELR QIEKDITSFL KKIIKDHQES LDRENPQDFI DMYLLHMEEE RK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a hydroxylase that metabolizes arachidonic acid, docosahexaenoic acid, and other long chain fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2U1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP2U1
Gene Accession No. Q7Z449
Gene Alias 8430436A10Rik; Cyp2u1; Cytochrome P450 2U1; cytochrome P450 family 2 subfamily U member 1; cytochrome P450, 2u1; cytochrome P450, family 2, subfamily U, polypeptide 1; P450TEC; spastic paraplegia 49; SPG49; SPG56
Gene Symbols CYP2U1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYP2U1. Recombinant protein control fragment (Product #RP-95661).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113612
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.