Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2U1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115085 Supplier Invitrogen™ Supplier No. PA5115085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 2U1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a hydroxylase that metabolizes arachidonic acid, docosahexaenoic acid, and other long chain fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2U1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CYP2U1
Gene Accession No. Q4V8D1, Q7Z449, Q9CX98
Gene Alias 8430436A10Rik; Cyp2u1; Cytochrome P450 2U1; cytochrome P450 family 2 subfamily U member 1; cytochrome P450, 2u1; cytochrome P450, family 2, subfamily U, polypeptide 1; P450TEC; spastic paraplegia 49; SPG49; SPG56
Gene Symbols CYP2U1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP2U1(Accession Q7Z449), corresponding to amino acid residues E286-S336.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113612, 310848, 71519
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.