Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP2W1 Mouse anti-Human, DyLight 680, Clone: V32-P5E5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250205FR
Description
CYP2W1 Monoclonal specifically detects CYP2W1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CYP2W1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYP2W1 | |
Ovalbumin-conjugated synthetic peptide - RYRLLPPPGV (C-terminal sequence) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
V32-P5E5 | |
Western Blot, ELISA, Immunohistochemistry | |
CYPIIW1, cytochrome P450 2W1, cytochrome P450, family 2, subfamily W, polypeptide 1, EC 1.14.14.-, EC 1.14.14.1, EC 1.14.14.5, FLJ20359, MGC34287 | |
Mouse | |
Protein A purified | |
RUO | |
54905 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction