Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYP2W1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550389 Supplier Invitrogen™ Supplier No. PA550389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CYP2W1 protein.

CYP2W1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2W1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Cyp2w1
Gene Accession No. Q8TAV3
Gene Alias CP2W1; CYP2W1; CYPIIW1; cytochrome P450 2W1; cytochrome P450 family 2 subfamily W member 1; cytochrome P450, family 2, subfamily w, polypeptide 1; Gm455
Gene Symbols Cyp2w1
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human cytochrome P450, family 2, subfamily W, polypeptide 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54905
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.