Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYP39A1 Antibody (M30P6D6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2502060.025ML
Description
CYP39A1 Monoclonal antibody specifically detects CYP39A1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
CYP39A1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Cytochrome P450 39A1, cytochrome P450, family 39, subfamily A, polypeptide 1, cytochrome P450, subfamily XXXIX (oxysterol 7 alpha-hydroxylase), polypeptide 1,24-hydroxycholesterol 7-alpha-hydroxylase, EC 1.14.13.99, hCYP39A1, oxysterol 7alpha-hydroxylase, Oxysterol 7-alpha-hydroxylase | |
Ovalbumin-conjugated synthetic peptide CRIEYKQRI (C-terminal sequence). | |
0.025 mL | |
Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
51302 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG3 κ |
Western Blot, ELISA, Immunohistochemistry | |
M30P6D6 | |
Unconjugated | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction